Learn More
Waters Corp Waters ACQUITY UPLC HSS C18 SB Column, 100 Å, 1.8 μm, 2.1 mm × 75 mm
Supplier:  Waters Corp 186005616
Encompass_Preferred
ACQUITY UPLC HSS C18 SB columns are designed for low pH separations involving complex mixtures of basic and non-basic compounds. These columns offer an alternative analyte selectivity compared to most modern, high-coverage, fully endcapped C18 stationary phases and are fully scalable and transferable to HPLC separations.
  • Increased retention for both polar and non-polar analytes.
  • Features thermally treated silica particles for enhanced mechanical strength and stability under UPLC system pressures.
  • Contributes to maximizing the selectivity space.
  • Facilitates various interaction types, including traditional hydrophobic, reversed-phase, dipole-dipole, aromatic, and hydrogen-bonding interactions.
Catalog No. 50-466-152