Learn More
Waters Corp UPLC Column, HSS SB C18, 100 Å, 1.8 μm, 2.1 × 100 mm
Supplier:  Waters Corp 176001561
Encompass_Preferred
This general-purpose, high-strength silica C18 column is mechanically tolerant of UPLC pressures. It is designed for reversed-phase separations, providing superior peak shape for basic compounds, increased retention for very polar compounds, and extremely long lifetimes. Its 100Å pore size and 1.8 μm particle size make it suitable for a wide range of applications, offering enhanced resolution, sensitivity, and speed.
  • Designed for reversed-phase separations with superior peak shape for basic compounds.
  • Increased retention for very polar compounds.
  • Extremely long lifetimes.
  • Mechanically tolerant of UPLC pressures.
  • Suitable for a wide range of applications.
  • Offers enhanced resolution, sensitivity, and speed.
Catalog No. 50-465-903