Learn More
Waters Corp Waters ACQUITY UPLC HSS C18 SB Column, 100Å, 1.8 μm, 1 mm x 150 mm
Supplier:  Waters Corp 176001558
Encompass_Preferred
ACQUITY UPLC HSS C18 SB columns are designed for low pH separations involving complex mixtures of basic and non-basic compounds. These columns offer an alternative analyte selectivity compared to most modern, high-coverage, fully endcapped C18 stationary phases. They are fully scalable and transferable to HPLC separations.
  • C18 chemistry
  • Reversed phase separation mode
  • 2 to 8 pH range
  • 45°C temperature limit
  • 18000 psi (1240 Bar) maximum pressure
  • 1.8 μm particle size
  • 100 Å pore size
  • 1 mm inner diameter
  • 150 mm length
Catalog No. 50-465-900