Learn More
Waters Corp ACQUITY UPLC HSS C18 SB Column, 100Å, 1.8 µm, 2.1 mm X 100 mm, 1/pk
Supplier:  Waters Corp 186004119
Encompass_Preferred
ACQUITY UPLC HSS C18 SB columns are used for low pH separations that contain complex mixtures of basic and non-basic compounds. ACQUITY HSS C18 SB columns give you alternative analyte selectivity compared to most modern, high coverage, fully endcapped C18 stationary phases. ACQUITY HSS C18 SB columns are fully scalable and transferable to HPLC separations.
Catalog No. 50-782-175