Learn More
Waters Corp ACQUITY UPLC Column, C18, 100Å, 1.8 μm, 1 × 150 mm
Supplier:  Waters Corp 176001123
Encompass_Preferred
The ACQUITY UPLC HSS C18 chemistry is an ultra-performance general purpose C18 bonded phase designed to deliver superior peak shape for bases, increased retention compared to ACQUITY UPLC BEH C18 columns, and exceptional column lifetimes at low pH. These ACQUITY HSS C18 columns are fully scalable and transferable to HPLC separations.
  • Superior peak shape for bases.
  • Increased retention.
  • Extremely long column lifetimes at low pH.
  • Fully scalable and transferable to HPLC separations.
Catalog No. 50-780-035