Learn More
Waters Corp ACQUITY UPLC HSS T3 Column, 100Å, 1.8 µm, 2.1 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186003540
Encompass_Preferred
The universal, silica-based bonded phase used for the ACQUITY HSS T3 sorbents is compatible with 100% aqueous mobile phase and should be your first choice when developing separations for polar and non-polar compounds. The ACQUITY HSS T3 UPLC columns exhibit ultra-low MS bleed and are fully scalable and transferable to HPLC separations.
Catalog No. 50-781-830