Learn More
Waters Corp ACQUITY UPLC Column, C18, 100 Å, 1.8 μm, 2.1 mm X 75 mm
Supplier:  Waters Corp 186005614
Encompass_Preferred
This column features a universal, silica-based bonded phase compatible with 100% aqueous mobile phase, suitable for developing separations for polar and non-polar compounds. It exhibits ultra-low MS bleed and is fully scalable and transferable to HPLC separations. The T3 sorbents eliminate the need for ion-pair reagents, while the HSS particle technology provides improved retentivity and alternate selectivity compared to hybrid particle technology.
  • Universal, silica-based bonded phase, 100% aqueous mobile phase compatible.
  • Separates polar and non-polar compounds effectively.
  • Ultra-low MS bleed; scalable to HPLC separations.
  • Low ligand density for enhanced retention of polar chemicals.
  • Eliminates ion-pair reagents for polar and hydrophobic compound retention.
  • HSS particle technology withstands UPLC pressures.
  • Improved retentivity and alternate selectivity.
Catalog No. 50-466-150