Learn More
Waters Corp Waters HSS T3 Column, 1.8 μm, 3 mm × 75 mm
Supplier:  Waters Corp 186005617
Encompass_Preferred
The ACQUITY UPLC HSS T3 Column is a universal, silica-based bonded phase compatible with 100% aqueous mobile phases for polar and non-polar compound separations. It exhibits ultra-low MS bleed and scales to HPLC. Incorporating MaxPeak High Performance Surface technology, it prevents unwanted analyte-surface interactions and maintains chemical stability under high backpressures due to its High-Strength Silica (HSS) base particle, reducing analysis time and cost.
  • Exhibits ultra-low MS bleed.
  • Scalable and transferable to HPLC separations.
  • MaxPeak High Performance Surface technology prevents analyte-surface interactions.
  • Reduces analysis time and cost per sample.
  • Available in over 50 column dimensions and configurations.
  • Compatible with pre-columns for enhanced protection and increased lifetime.
  • Improves sample throughput and chromatographic resolution.
Catalog No. 50-466-153