Learn More
Waters Corp ACQUITY UPLC M-Class Peptide Column, CSH C18, 1.7 μm
Supplier:  Waters Corp 186007514
Encompass_Preferred
ACQUITY UPLC M-Class Columns provide optimal nano- to micro-scale liquid chromatographic separation for biomarker discovery and peptide identification. Built to withstand high operating pressures (15 kpsi/1,000 bar), they ensure superior longevity. This column features CSH Technology with 130 Å pore, C18, 1.7 μm particles.
  • Optimal performance for nano- to micro-scale LC separation, biomarker discovery, and peptide identification.
  • High pressure tolerance (15000 psi / 1034 Bar) for superior longevity.
  • CSH particle technology enhances selectivity and peak shape, especially for basic compounds.
  • LC-MS compatible; minimal dependence on formic acid modifiers.
  • Increased peptide mass loads and fast equilibration for higher throughput.
  • Rigorous QC testing ensures reliability for peptide separation.
  • Manufactured in an ISO 9001 certified facility.
Catalog No. 50-239-0851