Learn More
Waters Corp ACQUITY UPLC M-Class Peptide BEH C18 Column, 1.7 μm, 75 μm x 100 mm
Supplier:  Waters Corp 186007481
Encompass_Preferred
ACQUITY UPLC M-Class columns are engineered for optimal performance in nano- to micro-scale liquid chromatographic separation, providing high sensitivity, resolution, and reproducibility for biomarker discovery and protein identification and characterization. These columns feature a novel frit technology, allowing them to withstand 15,000 psi operating pressure and manage pressure fluctuations during injection, ensuring superior longevity. They incorporate BEH Technology 130Å pore, C18, 1.7μm particles.
  • Each batch is specifically QC tested using a gradient separation of a tryptic digest of cytochrome c before being accepted as a Peptide separation column.
  • Proper connections are crucial to minimize peak tailing and ensure efficiency for maximum performance.
Catalog No. 50-239-2781