Learn More
Waters Corp Waters Column, CSH C18, 130Å 1.7 μm, 150 μm x 100 mm
Supplier: Waters Corp 186007480
ACQUITY UPLC M-Class Columns are designed for optimal performance in nano- to micro-scale liquid chromatographic separation for biomarker discovery and protein and peptide identification and characterization. These columns operate at 15 kpsi (1,000 bar) and manage pressure fluctuations, ensuring superior longevity.
- Incorporates CSH technology with 130 Å pore, C18, 1.7 μm particles.
- Delivers exceptional peak shape for basic compounds under low pH conditions.
- Minimal dependence on modifiers, making it suitable for LC-MS separations.
- Accepts greater peptide mass loads, improving detection of low-level constituents.
- Ensures fast column equilibration and pH switchover for increased sample throughput.
- Features a maximum pressure of 15000 psi (1034 bar) and a pH range of 1 to 10.
- Quality control tested using gradient separation of a tryptic digest of cytochrome c.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.