Learn More
Waters Corp ACQUITY UPLC Column, Peptide HSS T3, 1.8 μm, 1 mm X 100 mm
Supplier:  Waters Corp 186008752
Encompass_Preferred
These Peptide HSS T3 Columns are engineered for separations requiring silica-based selectivities or enhanced retention of hydrophilic peptides. Each batch of HSS T3, 100Å material undergoes rigorous quality control testing with a tryptic digest of cytochrome c to ensure stringent performance. Optimized for low dispersion micro-flow systems, these 1 mm ID columns are particularly effective for LC-MS methods.
  • Each batch of HSS T3, 100Å material is quality control tested with a gradient separation of a tryptic digest of cytochrome c and must pass stringent performance criteria.
  • 1 mm ID columns are recommended for use with low dispersion micro-flow systems, especially for LC-MS methods.
  • Suggested Use: Proteomics, Peptide Separations, Peptide Analysis, Micro-flow.
Catalog No. 50-239-2584