Learn More
Waters Corp ACQUITY UPLC Peptide HSS T3 C18 Column
Supplier:  Waters Corp 176003993
Encompass_Preferred
This column is designed for peptide separations and suitable for reversed-phase analysis of both hydrophilic and hydrophobic peptides.
  • Equipped with MaxPeak HPS technology to prevent unwanted analyte-surface interactions.
  • Provides ruggedness for reproducible data, attributed to the high-strength silica particle.
  • Offers a wide, usable pH range.
  • Aims to increase sample throughput and chromatographic resolution, reducing analysis time.
  • Includes a Cytochrome C digestion standard for immediate benchmarking and ongoing monitoring.
  • Recommended with the VanGuard pre-column for enhanced protection from fouling.
Catalog No. 50-239-2395