Learn More
Waters Corp ACQUITY UPLC Peptide HSS T3 C18 Column,  100Å , 1.8 µm, 2.1 x 150 mm, 3/pkg;Silica, USP Class:L1 , 2 - 8 Ph Range, Medium Silanol Activity, 230 m2/g, End Capped , Spherical , 11 % Carbon Load
Supplier:  Waters Corp 186008782
Encompass_Preferred
ACQUITY UPLC Peptide HSS T3 C18 Column,  100Å , 1.8 µm, 2.1 x 150 mm, 3/pkg;Silica, USP Class:L1 , 2 - 8 Ph Range, Medium Silanol Activity, 230 m2/g, End Capped , Spherical , 11 % Carbon Load
Catalog No. 50-146-1951