Learn More
Waters Corp Waters ACQUITY UPLC Column, Peptide HSS T3, 1.8 μm, 2.1 mm X 100 mm
Supplier:  Waters Corp 186008755
Encompass_Preferred
ACQUITY UPLC Peptide HSS T3 Columns are designed for separations requiring silica-based selectivities or increased retention of hydrophilic peptides. Each batch of HSS T3, 100Å material is QC tested with a gradient separation of a tryptic digest of cytochrome c. These 2.1 mm ID columns are recommended for low dispersion UPLC or UHPLC systems, especially for LC-MS methods.
  • Features low ligand density, ideal for reversed-phase liquid chromatography and enhanced retention of polar chemicals.
  • Utilizes HSS (High Strength Silica) particle technology for superior mechanical stability.
  • Offers higher retentivity and alternative selectivity compared to hybrid particle technology.
  • Employs Peptide Separation Technology (PST) for stable and well-developed peptide separations.
  • Generates crisp, symmetrical peaks across diverse peptide characteristics.
Catalog No. 50-146-1934