Learn More
Kyfora Bio Acridine orange 5g
Supplier:  Kyfora Bio 045395
Encompass
Acridine orange is a nucleic acid-selective fluorescent cationic dye useful for cell cycle determination It emits green fluorescence (525 nm) when bound to dsDNA and red fluorescence (650 nm) when bound to ssDNA or RNA
Catalog No. 50-002-85289