Learn More
Selleck Chemical LLC Acridine Orange hydrochloride
SDP
Supplier:  Selleck Chemical LLC E1838100MG
Encompass_Preferred
Acridine Orange hydrochloride is a cell-penetrable nucleic acid-selective fluorescent dye Acridine Orange hydrochloride produces orange fluorescence when it binds to ssDNA or RNA and green fluorescence when it binds to dsDNA (Ex 488 nM Em green fluorescence at 530 nm orange fluorescence at 640 nm)
Catalog No. 50-313-9055