Learn More
Medchemexpress LLC Acridone | 578-95-0 | 195.22 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW007771R
Encompass_Preferred
Acridone is an organic compound derived from the acridine skeleton. It exhibits a range of biological activities, including antibacterial, antimalarial, antiviral, and antineoplastic properties. It has been shown to inhibit the proliferation of human breast cancer cells in vitro.
  • Exhibits antibacterial properties
  • Shows antimalarial effects
  • Possesses antiviral activity
  • Demonstrates antineoplastic properties
  • Inhibits proliferation of human breast cancer cells
  • Blocks expression of ATP binding cassette subfamily G member 2 (ABCG2)
Catalog No. 50-004-49795