Learn More
Medchemexpress LLC Acriflavine hydrochloride | 69235-50-3 | 99.4% | 541.90 | C27H27Cl3N6 | 1 ML

Supplier: Medchemexpress LLC HYW08807510MM/1ML
Acriflavine hydrochloride is a fluorescent acridine dye and antiseptic agent used in biochemical and pharmacological research. It inhibits HIF-1 dimerization, interferes with MCT4-Basigin interactions, and shows activity against papain-like protease in antiviral studies.
- Formulation: 10 mM in DMSO, 1 mL
- Purity: 99.4%
- Cas number: 69235-50-3
- Molecular weight: 541.90
- Chemical formula: C27H27Cl3N6
- Appearance: brown to reddish brown solid; supplied as solution for small-volume use
- Solubility: soluble in DMSO (≈25 mg/mL); ultrasonic assistance may be required
- Storage: solid 4°C; in solvent -80°C (6 months) or -20°C (1 month)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.