Learn More
Medchemexpress LLC Acrivastine | 87848-99-5 | HY-B1510 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1510
Encompass_Preferred
Acrivastine is a short-acting histamine 1 receptor antagonist used for the treatment of allergic rhinitis. It is an effective and well-tolerated antihistamine, demonstrating superior efficacy to placebo and comparable results to other common antihistamines in treating chronic urticaria and seasonal allergic rhinitis.
  • Short-acting histamine 1 receptor antagonist
  • Effective in treating chronic urticaria and allergic rhinitis
  • Causes less drowsiness compared to clemastine
  • Reduces symptoms such as sneezing, runny nose, watery eyes, and itchy throat
  • Well tolerated
Catalog No. 50-004-14001