Learn More
TARGETMOL CHEMICALS INC ACRIZANIB 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T53735MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Acrizanib (LHA510) is a VEGFR-2 inhibitor with an IC50 of 17.4 nM for BaF3-VEGFR-2. purity: 98%
Catalog No. 50-257-9782