Learn More
Cytiva (Formerly Pall Lab) Acro 37 TF Vent Device with PTFE Membrane - 0.2 m, 37 mm (200/pkg)
Supplier:  Cytiva 4465
Encompass
Disposable, multipurpose filters for small volume venting, solvent, and gas filtration.-Hydrophobic membrane acts as a barrier to aqueous solutions and aerosols-Protect sensitive equipment or the laboratory environment-Help vent small volume culture vessels in in-line filtration of air and other gasses-Have Broad chemical compatibility for solvent filtrationComes with PTFE membrane and polypropylene housing. Membrane has 0.2m pore size and 37mm diameter with effective filtration area of 7.5cm2. Hose barb connectors are 6.49.5mm (1/43/8"). Maximum operating temperature: 100C (212F). Maximum operating pressure: 4.1bar (60psi), bidirectional. Nonsterile. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3046