Learn More
Cytiva (Formerly Pall Lab) Acro 50 Vent Devices with PTFE Membrane - 0.2 m, hose barb (72/pkg)
Supplier:  Cytiva 4250
Encompass
50 mm Filters with Superior Performance for Demanding ApplicationsReusable and autoclavablePTFE membrane and polypropylene housing have broad chemical resistanceLarge surface area provides greater throughput and high air flow rateAvailable with a variety of inlet and outlet connectors This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC0232694