Learn More
Cytiva AcroPrep™ 384-well Filter Plates, 100 µL - 1.0 µm glass fiber, long tips, natural (10/pkg)
Supplier:  Cytiva 5072N
Encompass
Ideal for Use With Low Sample Volume, High Throughput Sample Prep Applications-Plates are constructed from chemically-resistant, biologically-inert polypropylene with a clear polystyrene lid.-Robotic-friendly design has single-piece construction, designed in accordance with the ANSI/SBS X-2004 standards. Plates are stackable with and without lids.-Eliminates crosstalk between wells. Membranes are individually sealed in each well with a proprietary sealing process and each well has a fluid director.-A serialized barcode label allows the use of automated tracking systems and identifies the membrane type.-Available in a variety of membrane configurations to meet the requirements of numerous applications. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1530840