Learn More
Cytiva (Formerly Pall Lab) Acrodisc PSF Syringe Filter - GxF/Glass, 25 mm, polypropylene housing (50/pkg 200/cs)
Supplier:  Cytiva AP4523
Encompass
Acrodisc PSF Syringe Filter - GxF/Glass, 25 mm.Sample filtration helps protect sensitive parts of analytical instrumentation from particulate build-up, extending the life of chromatography columns and reducing system downtime. Acrodisc PSF syringe filters with glass fiber maximize throughput prefiltration of particulate-laden samples. They can be used alone or in series with another Acrodisc syringe filter.Acrodisc PSF syringe filters feature a large effective filtration area (3.9 cm2) for increased sample throughput, ideal for sample volumes of 150 mL. The syringe filters are designed to be fully compatible with automated robotic handling systems.They feature a GxF multi-layered glass fiber prefilter, providing two to four times the throughput of standard glass fiber prefilter devices, and allowing quick and easy filtration of your most difficult-to-filter samples. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3413