Learn More
Cytiva AcroPrep™ Advance 96-well Filter Plates for DNA Purification - 1 mL, DNA binding (5/pkg)
Supplier:  Cytiva 8132
Encompass
The Pall AcroPrep Advance 96-well Long Tip Filter Plate for Nucleic Acid Binding (NAB) offers researchers the flexibility to purify nucleic acids from a wide variety of starting material types: a single plate for multiple applications. The Pall NAB plate is a multipurpose plate providing flexibility in applications, reduced risk of cross contamination and smooth flow for sample processing.Maximum yields and quality of nucleic acid purification•Outlet tip geometry provides direct flow of samples into receiver plate without concerns of cross contamination•Silica-based quartz glass fiber media that allows efficient binding of DNA and RNA, while providing smooth flow and rapid processing of samples•Manufactured in accordance with ANSI/SBS x-2004 specifications for multi-well plates allowing entire DNA purification process to be performed on automated equipment This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3153