Learn More
Cytiva AcroPrep™ Advance 96-well Filter Plates for Solvent Filtration - 350 µL, Long Tip, 0.2 µm wwPTFE membrane (10/pkg)
Supplier:  Cytiva 8586
Encompass
Robust Chemical Resistance Provides Compatibility with Harsh Organics and Aqueous Solutions•Chemically resistant materials provide a stable platform to process samples in organic solvents•Inert materials of construction ensure complete recovery of samples and low non-specific binding•New well design results in faster, more uniform filtration rates across the plate and reduces hold-up volume•Innovative outlet tip geometry provides direct flow of samples into receiver plate without concerns of cross-contamination This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3161