Learn More
Waters Corp Acrodisc, Syringe Filter, PTFE, 4 mm, 0.45 µm, Non polar, 250/case
Supplier:  Waters Corp WAT200508
Encompass_Preferred
This PTFE membrane is a good choice for use with aggresive organic solvents
Catalog No. 50-786-055