Learn More
Cytiva (Formerly Pall Lab) Acrodisc PSF Syringe Filter with Supor Membrane - GxF/0.45 m, 25 mm (50/pkg 200/cs)
Supplier:  Cytiva AP4425
Encompass
Acrodisc PSF Syringe Filter with Supor Membrane - GxF/0.45 m, 25 mmSyringe filters with Supor (polyethersulfone) membrane are ideal for the filtration of aqueous and proteinaceous solutions in HPLC (0.45 m) and UHPLC (0.2 m) sample preparation. Sample filtration helps protect sensitive parts of analytical instrumentation from particulate build-up, extending chromatography column life and reducing system downtime.Acrodisc PSF Syringe Filters have a large effective filtration area (3.9 cm2) for increased sample throughput, ideal for sample volumes of 150 mL, and features a built-in GxF prefilter for use with difficult-to-filter solutions.The syringe filters are designed to be fully compatible with automated robotic handling systems. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1754442