Learn More
Cytiva (Formerly Pall Lab) AcroPak™ 20 Filters with Supor EKV Membrane - 0.2 m, gamma-irradiated (3/pkg)
Supplier:  Cytiva 12247
Encompass
Designed for Convenient Small-and Medium-Volume Filtration-Built-in MachV asymmetric prefilter layer for maximum flow and throughput performance-Disposable for reduced labor and associated costs-Significantly reduces the cost of repeat testing when scaling up your filtration system-Ideal for filtering aqueous solutions, cell culture media, and serum-Suitable for use with fluids containing dilute proteins, preservatives, or other critical components This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3196