Learn More
Cytiva (Formerly Pall Lab) AcroPak™ 200 Capsule with Supor Membrane - 0.8/0.2 m, gamma-irradiated, with filling bell (1/4 in. MNPT inlet, 1/4 - 1/2 in. stepped barb outlet) (3/pkg)
Supplier:  Cytiva 12093
Encompass
Efficient Processing of Liquid Volumes Up To 20 LSupor membrane has high flow rates and throughputs and is ideal for solutions where low protein binding is required. Not recommended for use with some ketones.Built-in prefilter extends filter life when particulate-laden solutions, such as serum-containing media, are processed.Upstream vent prevents vapor lock.Manufactured without the use of adhesives to minimize extractables.Sterilization by gamma irradiation eliminates potential contamination by EtO residuals. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1555007