Learn More
Cayman Chemical Acrvastn 10mg
SDP
Supplier:  Cayman Chemical 2402110 MG
Encompass_Preferred
A histamine H1 receptor antagonist (Ki = 10 nM in COS-7 cells expressing the human receptor); completely inhibits response to histamine in guinea pigs when administered at a dose of 1 mg/kg
Catalog No. 50-293-3185