Learn More
Alkali Scientific Acrylamide/bis Acrylamide, 29:1 Ratio Solution, 5 L
SDP
Supplier:  Alkali Scientific CRY008CP
Encompass_Preferred
Acrylamide/bis Acrylamide, 29:1 Ratio Solution, 5 L, is a high-volume solution designed for use in large-scale polyacrylamide gel electrophoresis (PAGE) experiments. The 29:1 ratio provides a more porous gel matrix, perfect for separating smaller proteins and nucleic acids. With a volume of 5 liters, this solution is ideal for high-throughput labs that require frequent and consistent electrophoresis experiments. The ready-to-use liquid solution saves preparation time and minimizes the risk of human error. It is commonly used in proteomics, molecular biology, and genomics research, where precise separation of small biomolecules is essential.
Catalog No. 50-006-60269