Learn More
Alkali Scientific ESR Tube Holder, Acrylic (PMMA), 10 Place Rack. 1/Pk, For Westergren ESR Pipettes
SDP
Supplier:  Alkali Scientific ESRRACK00
Encompass
The ESR Rack is manufactured from high clarity PMMA (polymethyl methacrylate) acrylic, offering excellent transparency, rigidity, and resistance to common laboratory chemicals. Designed for erythrocyte sedimentation rate (ESR) testing, each rack is precision molded to hold a single tube type securely in an upright position for accurate and reproducible results. Two models are available: the Westergren ESR Tube Rack measuring 305 (L) × 90 (W) × 180 (H) mm for standard Westergren tubes, and the 8 mm × 120 mm ESR Tube Rack measuring 202 (L) × 67 (W) × 91 (H) mm for 8 mm × 120 mm ESR tubes. Each rack features a 10 place configuration for consistent spacing, stability, and clear sample visibility. Smooth, easy to clean surfaces support hygienic handling and efficient laboratory workflows in clinical, hematology, and research settings.
Catalog No. 50-006-62331