Learn More
Omnimed Inc Acrylic Suture Rack - Triple Tier
SDP
Supplier:  Omnimed Inc 304102
Encompass_Preferred
Suture storage racks & carts keep sutures separated, organized, and easily accessible. Made of special impact resistant acrylic that’s more durable than traditional acrylic. Rack is front loading and rail free for quick restocking.
Catalog No. NC3956456