Learn More
Bioanalytical Systems Inc ACSF PERFUSION FLUID 25ML
SDP
Supplier:  Bioanalytical Systems Inc MD2400
Encompass
Premixed, sterile and ready to use. Commonly used as a perfusion fluid for CNS research applications (e.g., Interstitial fluid sampling, microdialysis). *Does not ship to Puerto Rico
Catalog No. NC1927309