Learn More
ABclonal Technology ACSL1 Rabbit PolymAb
SDP
Supplier:  ABclonal Technology A22737PM20UL
Encompass_Preferred
The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family Although differing in substrate specificity subcellular localization and tissue distribution all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters and thereby play a key role in lipid biosynthesis and fatty acid degradation Several transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-05192