Learn More
ABclonal Technology ACSM2A Rabbit pAb
SDP
Supplier:  ABclonal Technology A155631000UL
Encompass_Preferred
This gene encodes a mitochondrial acyl-coenzyme A synthetase that is specific for medium chain fatty acids. These enzymes catalyze fatty acid activation, the first step of fatty acid metabolism, through the transfer of acyl-CoA. These enzymes also participate in the glycine conjugation pathway in the detoxification of xenobiotics such as benzoate and ibuprofen. Expression levels of this gene in the kidney may be correlated with kidney function. This gene and its paralog ACSM2B (Gene ID 348158), both present on chromosome 16, likely arose from a chromosomal duplication event.
Catalog No. 50-001-08228