Learn More
Selleck Chemical LLC ACSS2 inhibitor 5mg 508186-14-9
SDP
Supplier:  Selleck Chemical LLC S85885MG
Encompass_Preferred
ACSS2 inhibitor is the most potent and specific inhibitor of acetate-dependent acetyl-CoA synthetase 2 (ACSS2). ACSS2 inhibitor also inhibits the respiratory syncytial virus (RSV). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1870606