Learn More
Medchemexpress LLC ACT-1004-1239 | 2178049-58-4 | 98.81% | C27H28F2N6O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY142617
Encompass_Preferred
ACT-1004-1239 is a potent, selective, and orally active CXCR7 antagonist with an IC50 value of 3.2 nM, developed for research purposes only. This compound promotes oligodendrocyte precursor cell differentiation and has shown promise in reducing disease severity in models of inflammatory demyelinating diseases.
  • Potent and selective CXCR7 antagonist
  • Orally active with an IC50 of 3.2 nM
  • Promotes oligodendrocyte precursor cell (OPC) differentiation
  • Increases plasma CXCL12 concentration in mice
  • Reduces disease severity in MOG-induced experimental autoimmune encephalomyelitis (EAE) model
  • Increases myelination in Cuprizone-induced demyelination model mice
Catalog No. 50-003-56712