Learn More
Selleck Chemical LLC ACT-1016-0707-E1941-1G
SDP
Supplier:  Selleck Chemical LLC E19411G
Encompass_Preferred
ACT-1016-0707 (LPA1 receptor antagonist 2) is an orally active and selective antagonist of LPA1 Receptor with IC50 of 3 1 nM for hLPAR1 in tango assay It has the potential to treat fibrotic diseases
Catalog No. 50-313-7679