Learn More
Medchemexpress LLC HY-124071 5mg Medchemexpress, ACT-389949 CAS:1258417-54-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1240715MG
Encompass_Preferred
Medchemexpress, HY-124071 5mg ACT-389949 CAS:1258417-54-7 ACT-389949 is a first-in-class, potent and selective and agonist of formyl peptide receptor type 2 (FPR2)/Lipoxin A4 receptor (ALX), with an EC50 of 3 nM for FPR2/ALX internalization into monocytes. ACT-389949 has potential for the treatment of inflammatory disorders[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9980