Learn More
Medchemexpress LLC Act-462206 | 1361321-96-1 | MFCD28502270 | 99.5% | 388.48 | C20H24N2O4S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10183425MG
Encompass_Preferred
ACT-462206 is an orally active, brain-penetrant dual orexin (OX1/OX2) receptor antagonist used in neuroscience research into sleep, anxiety, and addiction. The compound shows low-nanomolar activity at orexin receptors and is supplied as a high-purity solid or solution format for laboratory use (research use only).
  • Dual orexin receptor antagonist with low-nanomolar potency (OX1 and OX2)
  • Brain-penetrant properties suitable for CNS pharmacology studies
  • High purity (~99.5%) for reliable biochemical and pharmacological testing
  • Soluble in DMSO at ≥100 mg/mL for in vitro preparations
  • Available in research-scale solid and solution formats
  • Stored and shipped under recommended conditions to preserve stability
  • For research use only, not for human or clinical use
Catalog No. 50-002-48621