Learn More
Medchemexpress LLC Act-660602 | 1646267-59-5 | C20H20F6N8OS | 100 MG
SDP
Supplier:  Medchemexpress LLC HY151096100MG
Encompass_Preferred
ACT-660602 is an orally active antagonist of chemokine receptor (CXCR3) with an IC50 value of 204 nM. It inhibits T-cell migration and has shown efficacy in an acute lung injury model. This compound can be used for autoimmune diseases research.
  • Orally active antagonist of chemokine receptor (CXCR3)
  • Inhibits T-cell migration
  • Demonstrates efficacy in an acute lung injury model
  • Suitable for autoimmune diseases research
  • Exhibits selectivity for CXCR3 over hERG (IC50 18 μM)
  • Displays non-competitive binding to CXCL10 and CXCL11
Catalog No. 50-003-75847