Learn More
Medchemexpress LLC HY-112723 10mg Medchemexpress, ACT-709478 CAS:1838651-58-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11272310MG
Encompass_Preferred
Medchemexpress, HY-112723 10mg ACT-709478 CAS:1838651-58-3 ACT-709478 is a potent, selective, orally active, and brain penetrating T-type calcium channel blocker. ACT-709478 is used in the research of generalized epilepsies. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9158