Learn More
Medchemexpress LLC ACTH (1-17) TFA (α1-17-ACTH TFA) | 99.79% | 2207.43 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1545A10MG
Encompass_Preferred
ACTH (1-17) TFA is an adrenocorticotropin analogue and a potent human melanocortin 1 (MC1) receptor agonist with a Ki of 0.21 nM. It exhibits high affinity for the hMC1R and can induce a slight increase in growth hormone secretion in rat pituitary cultures. In vivo studies show it inhibits DNA labeling.
  • Potent human melanocortin 1 (MC1) receptor agonist
  • High purity of 99.79%
  • Detailed information on solubility and stock solution preparation
  • Supported by published research on its biological activity
  • For research use only
Catalog No. 50-004-24317