Learn More
Bachem ACTH (1-39) (mouse, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4034845.1000
Encompass_Preferred
ACTH (1-39) (mouse, rat) Trifluoroacetate, H-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-Asn-Val-Ala-Glu-Asn-Glu-Ser-Ala-Glu-Ala-Phe-Pro-Leu-Glu-Phe-OH, 77465-10-2, C210H315N57O57S, Mr 4582.23, 1mg. ACTH (1–39) is a member of the melanocortin receptor agonists, which stimulates the adrenal cortex and the secretion of glucocorticoids such as cortisol.
Catalog No. 50-259-914