Learn More
Medchemexpress LLC ACTH (34-39) | 69454-10-0 | 99.9% | 722.83 | 1 MG

Supplier: Medchemexpress LLC HYP17391MG
ACTH (34-39) is an adrenocorticotropic hormone fragment primarily used in the study of stress-related sleep disorders. This peptide targets the Melanocortin Receptor, playing a role in GPCR/G Protein pathways and neuronal signaling. It exhibits weak cross-reactivity with CLIP antibodies and is for research use only.
- Adrenocorticotropic hormone fragment
- Used in stress-related sleep disorders research
- Sequence: Ala-Phe-Pro-Leu-Glu-Phe (AFPLEF)
- Targets Melanocortin Receptor
- Involved in GPCR/G Protein pathway and Neuronal Signaling
- Weak cross-reactivity with CLIP antibodies (10%)
- Store sealed, away from moisture
- Powder storage: -80°C for 2 years, -20°C for 1 year
- In solvent storage: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.