Learn More
Medchemexpress LLC ACTH (7-38) (human) | 68563-24-6 | 99.6% | 3659.11 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP487410MG
Encompass_Preferred
ACTH (7-38) (human) is the 7-38 fragment of human ACTH (1-39). Human ACTH (1-39), known as a corticotropin inhibitory peptide (CIP), is an antagonist of the ACTH receptor and has no corticosteroid activity.
  • 7-38 fragment of human ACTH (1-39)
  • Antagonist of the ACTH receptor
  • No corticosteroid activity
  • Used in research related to metabolic or endocrine disease
  • Inhibitor of Melanocortin Receptor signaling pathway
Catalog No. 50-003-98225