Learn More
Cayman Chemical ACTH18-39humn 2.5mg
SDP
Supplier:  Cayman Chemical 272742.5 MG
Encompass_Preferred
A C-terminal peptide fragment of ACTH; produced via processing of ACTH in the intermediate lobe of the pituitary gland; increases cumulative food intake in fasted, but not fed, rats at 2.5 nmol/animal, intracerebroventricularly
Catalog No. 50-293-8276